missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Lithium Nitrate AR Granular, Macron Fine Chemicals™

macron

Chemical Identifiers

CAS 7790-69-4
Physical Form Granular
Molecular Formula LiNO3
Formula Weight 68.95
Chemical Name or Material Lithium Nitrate
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
MK5872125 Avantor Performance
587202
POLYSTORMOR 125g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
MK5872212 Avantor Performance
587206
Poly 2.5kg N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
MK5872500 Avantor Performance
587204
POLYSTORMOR 500g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

7790-69-4
LiNO3
Lithium Nitrate
Granular
68.95

Specifications

7790-69-4
LiNO3
POLYSTORMOR
Lithium Nitrate
Granular
68.95
125g