missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Lithium Nitrate AR Granular, Macron Fine Chemicals™

macron

Manufacturer:  Avantor Performance 587204

 View more versions of this product

Catalog No. MK5872500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Lithium Nitrate
LiNO3
IIPYXGDZVMZOAP-UHFFFAOYSA-N
lithium;nitrate
10129889
68.95
POLYSTORMOR
7790-69-4
lithium nitrate, nitric acid, lithium salt, lithium nitrate, anhydrous, lithium 1+ ion nitronate, ksc492i7j, lithium nitrate 3-hydrate, lithium 1+ ion nitrate, lithium nitrate, anhydrous 25g, lithium nitrate, ionization buffer solution, specpure
[Li+].[N+](=O)([O-])[O-]
68.944
CHEBI:63315
Granular
500g

Chemical Identifiers

7790-69-4
68.944
lithium nitrate, nitric acid, lithium salt, lithium nitrate, anhydrous, lithium 1+ ion nitronate, ksc492i7j, lithium nitrate 3-hydrate, lithium 1+ ion nitrate, lithium nitrate, anhydrous 25g, lithium nitrate, ionization buffer solution, specpure
CHEBI:63315
[Li+].[N+](=O)([O-])[O-]
LiNO3
IIPYXGDZVMZOAP-UHFFFAOYSA-N
10129889
lithium;nitrate
Documents