Please login to your online account to display your discounted pricing

Lithium Nitrate AR Granular, Macron Fine Chemicals™


macron

Quantity 2.5kg
Packaging Poly

Manufacturer: avantor performance  587206

 View more versions of this product

Catalog No. MK5872212

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 68.95
Quantity 2.5kg
Physical Form Granular
Molecular Formula LiNO3
CAS 7790-69-4
Chemical Name or Material Lithium Nitrate
Packaging Poly

Specifications

Chemical Name or Material Lithium Nitrate
CAS 7790-69-4
Physical Form Granular
Molecular Formula LiNO3
Formula Weight 68.95