Please login to your online account to display your discounted pricing

Lithium Nitrate AR Granular, Macron Fine Chemicals™


macron

Quantity 125g
Packaging POLYSTORMOR

Manufacturer: avantor performance  587202

 View more versions of this product

Catalog No. MK5872125

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 68.95
Physical Form Granular
Chemical Name or Material Lithium Nitrate
Packaging POLYSTORMOR
Molecular Formula LiNO3
CAS 7790-69-4
Quantity 125g

Specifications

Chemical Name or Material Lithium Nitrate
CAS 7790-69-4
Physical Form Granular
Molecular Formula LiNO3
Formula Weight 68.95