missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Lithium nitrate, anhydrous, 99%, Alfa Aesar™

Used in Ceramics, Pyrotechnics, Salt Baths

Manufacturer:  Alfa Aesar 1340514

 View more versions of this product

Catalog No. AA1340514



Specifications

Specifications

Lithium nitrate, anhydrous
99%
MFCD00011094
lithium nitrate, nitric acid, lithium salt, lithium nitrate, anhydrous, lithium 1+ ion nitronate, ksc492i7j, lithium nitrate 3-hydrate, lithium 1+ ion nitrate, lithium nitrate, anhydrous 25g, lithium nitrate, ionization buffer solution, specpure
[Li+].[N+](=O)([O-])[O-]
68.944
CHEBI:63315
Crystals
Odorless
25g
7790-69-4
LiNO3
White crystalline
IIPYXGDZVMZOAP-UHFFFAOYSA-N
lithium;nitrate
10129889
68.95
2.38g/cm3 at 20°C
White

Chemical Identifiers

7790-69-4
68.944
IIPYXGDZVMZOAP-UHFFFAOYSA-N
10129889
lithium;nitrate
LiNO3
MFCD00011094
lithium nitrate, nitric acid, lithium salt, lithium nitrate, anhydrous, lithium 1+ ion nitronate, ksc492i7j, lithium nitrate 3-hydrate, lithium 1+ ion nitrate, lithium nitrate, anhydrous 25g, lithium nitrate, ionization buffer solution, specpure
CHEBI:63315
[Li+].[N+](=O)([O-])[O-]
Safety and Handling

Safety and Handling

H272-H315-H319-H335: May intensify fire; oxidizer. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.

DOTInformation : DOT Class: 5.1, Packaging Group: III

EINECSNumber : 232-218-9

Documents