Learn More
Lithium nitrate, anhydrous, 99.98% (metals basis), Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 01098509
Description
Lithium nitrate, anhydrous is used in the manufacture of red-colored fireworks and flares. has been proposed as a medium to store heat collected from the sun for cooking. A Fresnel lens would be used to melt solid lithium nitrate, which would then function as a 'solar battery', allowing heat to be redistributed later by convection
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
600°C (decomposition) | |
264°C | |
7790-69-4 | |
10 g | |
LiNO3 | |
UN2722 | |
lithium nitrate, nitric acid, lithium salt, lithium nitrate, anhydrous, lithium 1+ ion nitronate, ksc492i7j, lithium nitrate 3-hydrate, lithium 1+ ion nitrate, lithium nitrate, anhydrous 25g, lithium nitrate, ionization buffer solution, specpure | |
IIPYXGDZVMZOAP-UHFFFAOYSA-N | |
lithium;nitrate | |
10129889 | |
68.95 | |
Odorless | |
2.38 g/mL |
Lithium nitrate | |
Anhydrous | |
Crystalline | |
(metals basis) | |
MFCD00011094 | |
14,5536 | |
Soluble in water. Also soluble in ethanol,methanol,pyridine,ammonia and acetone. | |
[Li+].[O-][N+]([O-])=O | |
68.94 | |
CHEBI:63315 | |
99.98% | |
Hygroscopic |
Chemical Identifiers
7790-69-4 | |
68.94 | |
IIPYXGDZVMZOAP-UHFFFAOYSA-N | |
10129889 | |
lithium;nitrate |
LiNO3 | |
MFCD00011094 | |
lithium nitrate, nitric acid, lithium salt, lithium nitrate, anhydrous, lithium 1+ ion nitronate, ksc492i7j, lithium nitrate 3-hydrate, lithium 1+ ion nitrate, lithium nitrate, anhydrous 25g, lithium nitrate, ionization buffer solution, specpure | |
CHEBI:63315 | |
[Li+].[O-][N+]([O-])=O |
Safety and Handling
GHS H Statement
H272-H315-H319-H335
May intensify fire
oxidizer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P220-P221-P264b-P270-P280-P301+P312-P305+P351+P338-P330-P370+P378q-P501c
H272-H302-H319
DOTInformation : Transport Hazard Class: 5.1; Packing Group: III; Proper Shipping Name: LITHIUM NITRATE
EINECSNumber : 232-218-9
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only