Learn More
Lithium nitrate, 99.999%, (trace metal basis), Thermo Scientific™
Supplier: Thermo Scientific Chemicals 212540250
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
for ultra trace analysisSpecifications
Lithium nitrate | |
25g | |
LiNO3 | |
15,5592 | |
Solubility in water: 90g/L (28°C). Other solubilities: soluble in alcohol | |
[Li+].[N+](=O)([O-])[O-] | |
68.95 | |
CHEBI:63315 | |
99.999% | |
Glass bottle | |
2.3800g/mL |
7790-69-4 | |
99.999% | |
LiNO3 | |
lithium nitrate, nitric acid, lithium salt, lithium nitrate, anhydrous, lithium 1+ ion nitronate, ksc492i7j, lithium nitrate 3-hydrate, lithium 1+ ion nitrate, lithium nitrate, anhydrous 25g, lithium nitrate, ionization buffer solution, specpure | |
IIPYXGDZVMZOAP-UHFFFAOYSA-N | |
lithium;nitrate | |
10129889 | |
68.95 | |
Trace Metal Basis | |
2.38 |
Safety and Handling
GHS H Statement
Harmful if swallowed.
May intensify fire; oxidizer.
Causes serious eye irritation.
GHS P Statement
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove conta
GHS Signal Word: Warning
EINECSNumber : 232-218-9
RUO â Research Use Only