missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Lithium niobium oxide, Puratronic™, 99.998% (metals basis excluding Ta), Ta <50ppm, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 01074114
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Lithium niobium oxide | |
12031-63-9 | |
25 g | |
LiNbO3 | |
Lithium niobate | |
[Li+].[O-][Nb](=O)=O | |
147.84 | |
147.85 | |
Puratronic™ |
∼1275°C | |
Powder | |
(metals basis excluding Ta), Ta <50ppm | |
MFCD00011080 | |
GQYHUHYESMUTHG-UHFFFAOYSA-N | |
lithium;oxido(dioxo)niobium | |
23665650 | |
99.998% | |
4.3 g/mL |
Chemical Identifiers
12031-63-9 | |
147.84 | |
GQYHUHYESMUTHG-UHFFFAOYSA-N | |
23665650 | |
[Li+].[O-][Nb](=O)=O |
LiNbO3 | |
MFCD00011080 | |
Lithium niobate | |
lithium;oxido(dioxo)niobium |
Safety and Handling
EINECSNumber : 234-755-4
RTECSNumber : QT9800000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only