missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Lithium niobium oxide, Puratronic™, 99.998% (metals basis excluding Ta), Ta <50ppm

Manufacturer:  Alfa Aesar™ 1074122

 View more versions of this product

Catalog No. AA1074122



Specifications

Specifications

Lithium niobium oxide
(metals basis excluding Ta), Ta <50ppm
MFCD00011080
GQYHUHYESMUTHG-UHFFFAOYSA-N
lithium;oxido(dioxo)niobium
23665650
Powder
Puratronic™
~1275°C
12031-63-9
LiNbO3
Lithium niobate
[Li+].[O-][Nb](=O)=O
147.843
147.85
99.998%
4.3g/mL
100g

Chemical Identifiers

12031-63-9
147.843
GQYHUHYESMUTHG-UHFFFAOYSA-N
23665650
[Li+].[O-][Nb](=O)=O
LiNbO3
MFCD00011080
Lithium niobate
lithium;oxido(dioxo)niobium
Safety and Handling

Safety and Handling

RTECSNumber : QT9800000

TSCA : TSCA

Documents