Please login to your online account to display your discounted pricing

Alfa Aesar™ Lithium niobium oxide, 99.99%

$32.64 - $595.70

Chemical Identifiers

Chemical Name or Material Lithium niobium oxide
CAS 12031-63-9
Molecular Formula LiNbO3
Formula Weight 147.85
MDL Number MFCD00011080
Synonym Lithium niobate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1780506

 
alfa aesar™
A1780506*
5g Each for $32.64

AAA1780530

 
alfa aesar™
A1780530
250g Each for $595.70

AAA1780518

 
alfa aesar™
A1780518
50g Each for $149.00
Description & Specifications

Specifications

Formula Weight 147.85
Chemical Name or Material Lithium niobium oxide
TSCA TSCA
Odor Odorless
Molecular Formula LiNbO3
MDL Number MFCD00011080
Synonym Lithium niobate
Melting Point ≈1275°C
Quantity 5g
RTECS Number QT9800000
Assay 99.99%
CAS 12031-63-9

Applications
It is employed in the manufacture of optical waveguides and surface acoustic wave devices, Lithium niobate (CAS NO.12031-63-9) is used extensively in the telecoms market, e.g. in mobile telephones and optical modulators. It can also find its uses in laser frequency doubling, nonlinear optics, Pockels cells, optical parametric oscillators, Q-switching devices for lasers, other acousto-optic devices, optical switches for gigahertz frequencies.

Solubility
Soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed in a cool, dry and well ventilated place.