missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Lithium niobium isopropoxide, 99.9% (metals basis)

Manufacturer:  Alfa Aesar™ 4445106

 View more versions of this product

Catalog No. AA4445106



Description

Description

  • Packaged under Argon
Specifications

Specifications

Lithium niobium isopropoxide
140°C (0.2mmHg)
21864-21-1
LiNb(OC3H7)6
UN2925
Solid,Packaged under Argon
Moisture sensitive
5g
(metals basis)
MFCD00210626
454.38
99.9%

Chemical Identifiers

21864-21-1
LiNb(OC3H7)6
MFCD00210626
Safety and Handling

Safety and Handling

GHS H Statement
H314-H228
Causes severe skin burns and eye damage.
Flammable solid.

P210-P240-P241-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c

H228-H314-H335

DOTInformation : Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: FLAMMABLE SOLIDS, CORROSIVE, ORGANIC, N.O.S.

TSCA : No

storageNote1 :

  • Packaged under Argon

Recommended Storage : Ambient temperatures

Documents