missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Lithium molybdenum oxide, 99+%

Manufacturer:  Alfa Aesar™ 1342736

 View more versions of this product

Catalog No. AA1342736



Specifications

Specifications

Lithium molybdenum oxide
Li2MoO4
lithium molybdate, dilithium 1+ ion molybdate, dilithium dioxido dioxo molybdenum, dilithotab molybdate, 2li.moo4
[Li+].[Li+].[O-][Mo](=O)(=O)[O-]
173.826
173.83
≥99%
Odorless
White
500g
13568-40-6
MFCD00016178
NMHMDUCCVHOJQI-UHFFFAOYSA-N
dilithium;dioxido(dioxo)molybdenum
6093689
Powder
2.66g/mL
Hygroscopic
705°C

Chemical Identifiers

13568-40-6
173.826
NMHMDUCCVHOJQI-UHFFFAOYSA-N
6093689
[Li+].[Li+].[O-][Mo](=O)(=O)[O-]
Li2MoO4
MFCD00016178
lithium molybdate, dilithium 1+ ion molybdate, dilithium dioxido dioxo molybdenum, dilithotab molybdate, 2li.moo4
dilithium;dioxido(dioxo)molybdenum
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA

Documents