Learn More
Lithium methoxide, pure, 2.2M (10 wt%) solution in methanol, AcroSeal™, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 336751000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Lithium methoxide | |
10% Solution in Methanol | |
CH3LiO | |
MFCD00036357 | |
lithium methoxide, lithium methanolate, methanol, lithium salt, lithium methylate, liome, methanol, lithium salt 1:1, liom, lithium methylat, lithotab methoxide, lioch3 | |
[Li+].C[O-] | |
37.97 | |
37.97 | |
Pure | |
100 mL |
Colorless | |
865-34-9,67-56-1 | |
CH3OLi | |
0.8500g/mL | |
JILPJDVXYVTZDQ-UHFFFAOYSA-N | |
lithium;methanolate | |
4177230 | |
≥10% | |
AcroSeal™ Glass Bottle in Tin Can | |
Liquid |
Chemical Identifiers
865-34-9 | |
37.97 | |
JILPJDVXYVTZDQ-UHFFFAOYSA-N | |
4177230 | |
[Li+].C[O-] |
CH3LiO | |
MFCD00036357 | |
lithium methoxide, lithium methanolate, methanol, lithium salt, lithium methylate, liome, methanol, lithium salt 1:1, liom, lithium methylat, lithotab methoxide, lioch3 | |
lithium;methanolate |
Safety and Handling
GHS P Statement
Causes damage to organs.
Toxic if swallowed.
Causes severe skin burns and eye damage.
Toxic if inhaled.
Toxic in contact with skin.
Highly flammable liquid and vapor.
Self-heating; may catch fire.
Reacts violently with water.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF exposed: Call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Danger
EINECSNumber : 212-737-7
TSCA : TSCA
RUO – Research Use Only