Please login to your online account to display your discounted pricing

Lithium methoxide, pure, 2.2M (10 wt%) solution in methanol, AcroSeal™, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Lithium methoxide
Name Note 10% Solution in Methanol
CAS 865-34-9
CAS 67-56-1
Purity Grade Pure
Assay ≥10%
Molecular Formula CH3LiO
Linear Formula CH3OLi
Formula Weight 37.97
MDL Number MFCD00036357
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC336758000

  SDS
thermo fisher scientific
AC336758000
AcroSeal™ Glass Bottle 800mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC336751000

  SDS
thermo fisher scientific
336751000
AcroSeal™ Glass Bottle in Tin Can 100mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥10%
Physical Form Liquid
Formula Weight 37.97
Quantity 800mL
Color Clear colorless
Packaging AcroSeal™ Glass Bottle
TSCA TSCA
Density 0.8500g/mL
EINECS Number 212-737-7
Purity Grade Pure
Chemical Name or Material Lithium methoxide
Name Note 10% Solution in Methanol
Molecular Formula CH3LiO
Linear Formula CH3OLi
MDL Number MFCD00036357
CAS 865-34-9
Health Hazard 1 Danger
Health Hazard 2 GHS P Statement
Causes damage to organs.
Toxic if swallowed.
Causes severe skin burns and eye damage.
Toxic if inhaled.
Toxic in contact with skin.
Highly flammable liquid and vapor.
Self-heating; may catch fire.
Reacts violently with water.
CAS 67-56-1
Health Hazard 3 GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF exposed: Call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

Danger

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF exposed: Call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS P Statement
Causes damage to organs.
Toxic if swallowed.
Causes severe skin burns and eye damage.
Toxic if inhaled.
Toxic in contact with skin.
Highly flammable liquid and vapor.
Self-heating; may catch fire.
Reacts violently with water.