missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Lithium methoxide, 98+%

Manufacturer:  Alfa Aesar™ 4412914

 View more versions of this product

Catalog No. AA4412914



Specifications

Specifications

Lithium methoxide
White
25g
CH3LiO
MFCD00036357
3552389
Soluble in methanol.
[Li+].C[O-]
37.974
37.98
≥98%
Air and moisture sensitive
>500°C (decomposition)
865-34-9
LiOCH3
UN3206
lioch3
JILPJDVXYVTZDQ-UHFFFAOYSA-N
lithium;methanolate
4177230
Powder

Chemical Identifiers

865-34-9
37.974
JILPJDVXYVTZDQ-UHFFFAOYSA-N
4177230
[Li+].C[O-]
CH3LiO
MFCD00036357
lioch3
lithium;methanolate
Safety and Handling

Safety and Handling

GHS H Statement
H251-H314
Self-heating: may catch fire.
Causes severe skin burns and eye damage.

P235+P410-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-X

DOTInformation : Transport Hazard Class: 4.2; Packing Group: II; Proper Shipping Name: ALKALI METAL ALCOHOLATES, SELF-HEATING, CORROSIVE, N.O.S.

EINECSNumber : 212-737-7

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents