Please login to your online account to display your discounted pricing

Lithium metasilicate, 99.5% (metals basis), Alfa Aesar™

Chemical Identifiers

Chemical Name or Material Lithium metasilicate
CAS 10102-24-6
Physical Form White powder
Assay Percent Range 99.5% (Metal basis)
Molecular Formula Li2O3Si
Formula Weight 89.94g/mol
MDL Number MFCD00016179
Synonym Lithium silicate (meta)
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4065622

 
alfa aesar
40656-22
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Appearance White powder
Density 2.52g/cm3 at 20°C
Odor Odorless
Color White
Quantity 100g
Chemical Name or Material Lithium metasilicate
CAS 10102-24-6
Physical Form White powder
Assay Percent Range 99.5% (Metal basis)
Molecular Formula Li2O3Si
Formula Weight 89.94g/mol
MDL Number MFCD00016179
Synonym Lithium silicate (meta)
Health Hazard 1 H315-H319-H335: Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.
EINECS Number 233-270-5
Safety and Handling

H315-H319-H335: Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.