Please login to your online account to display your discounted pricing

Alfa Aesar™ Lithium metagallate, 99%

Chemical Identifiers

Chemical Name or Material Lithium metagallate
CAS 944251-30-3
Linear Formula LiGaO2
Formula Weight 108.66
MDL Number MFCD00210623
Synonym Lithium gallate(III); Gallium(III) lithium oxide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3937006

 
alfa aesar™
3937006
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA3937014

 
alfa aesar™
3937014
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 108.66
Chemical Name or Material Lithium metagallate
Physical Form Powder
Assay 99%
Linear Formula LiGaO2
MDL Number MFCD00210623
CAS 944251-30-3
Synonym Lithium gallate(III); Gallium(III) lithium oxide
Color White
Melting Point >400°C
Quantity 5g