missing translation for 'onlineSavingsMsg'
Learn More

Lithium metaborate, dried, 99%

CAS: 13453-69-5 | B4H14Li2O7 | 183.23 g/mol

$73.42 - $697.30

Chemical Identifiers

CAS 13453-69-5
Molecular Formula B4H14Li2O7
Molecular Weight (g/mol) 183.23
MDL Number MFCD00011089
InChI Key DPYKRXYVXDYLEY-UHFFFAOYSA-N
Synonym lithium metaborate, boric acid hbo2 , lithium salt, lithium metaborate, anhydrous, lithium oxido oxo borane, bo2.li, boric acid hbo2 , lithium salt 1:1, lithium metaborate,anhydrous, lithium metaborate, puratronic, lithotab boron oxide hydroxide, lithium metaborate, acs
PubChem CID 123308
IUPAC Name lithium;oxido(oxo)borane
SMILES [Li].[Li].[B].[B].[B].[B].O.O.O.O.O.O.O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1614514
View Documents
Thermo Scientific Chemicals
A1614514
25 g
Each for $73.42
Only null left
 
AAA1614522
View Documents
Thermo Scientific Chemicals
A1614522
100 g
Each for $208.30
Only null left
 
AAA1614536
View Documents
Thermo Scientific Chemicals
A1614536
500 g
Each for $697.30
Only null left
 
Description

Description

Lithium metaborate is an effective flux for silicates and other rock-forming minerals. An improved lithium metaborate fusion technique for inductively coupled plasma mass spectrometric analysis of felsic rocks is proposed. The determination of fluorine in silicate materials with a fluoride ion-selective electrode following fusion with lithium metaborate is described.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Lithium metaborate is an effective flux for silicates and other rock-forming minerals. An improved lithium metaborate fusion technique for inductively coupled plasma mass spectrometric analysis of felsic rocks is proposed. The determination of fluorine in silicate materials with a fluoride ion-selective electrode following fusion with lithium metaborate is described.

Solubility
Soluble in water(27.5g/L).

Notes
Hygroscopic. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, water, moisture, alkali metals, acid anhydrides, acids.
Specifications

Chemical Identifiers

13453-69-5
183.23
DPYKRXYVXDYLEY-UHFFFAOYSA-N
123308
[Li].[Li].[B].[B].[B].[B].O.O.O.O.O.O.O
B4H14Li2O7
MFCD00011089
lithium metaborate, boric acid hbo2 , lithium salt, lithium metaborate, anhydrous, lithium oxido oxo borane, bo2.li, boric acid hbo2 , lithium salt 1:1, lithium metaborate,anhydrous, lithium metaborate, puratronic, lithotab boron oxide hydroxide, lithium metaborate, acs
lithium;oxido(oxo)borane

Specifications

845°C
25 g
LiBO2
lithium metaborate, boric acid hbo2 , lithium salt, lithium metaborate, anhydrous, lithium oxido oxo borane, bo2.li, boric acid hbo2 , lithium salt 1:1, lithium metaborate,anhydrous, lithium metaborate, puratronic, lithotab boron oxide hydroxide, lithium metaborate, acs
DPYKRXYVXDYLEY-UHFFFAOYSA-N
lithium;oxido(oxo)borane
123308
99%
Hygroscopic
Lithium metaborate, dried
13453-69-5
B4H14Li2O7
MFCD00011089
Soluble in water(27.5g/L).
[Li].[Li].[B].[B].[B].[B].O.O.O.O.O.O.O
183.23
49.75
Odorless
1.397
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

EINECSNumber : 236-631-5

TSCA : Yes

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only