missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Lithium metaborate, anhydrous, tech.

Manufacturer:  Alfa Aesar™ 3257636

 View more versions of this product

Catalog No. AA3257636



Specifications

Specifications

Lithium metaborate
13453-69-5
LiBO2
lithium metaborate, boric acid hbo2 , lithium salt, lithium metaborate, anhydrous, lithium oxido oxo borane, bo2.li, boric acid hbo2 , lithium salt 1:1, lithium metaborate,anhydrous, lithium metaborate, puratronic, lithotab boron oxide hydroxide, lithium metaborate, acs
[Li+].B(=O)[O-]
49.748
49.75
Technical
Odorless
White
500g
Anhydrous
BLiO2
MFCD00011089
HZRMTWQRDMYLNW-UHFFFAOYSA-N
lithium;oxido(oxo)borane
123308
Powder
1.397g/mL
Hygroscopic
845°C

Chemical Identifiers

13453-69-5
49.748
HZRMTWQRDMYLNW-UHFFFAOYSA-N
123308
[Li+].B(=O)[O-]
BLiO2
MFCD00011089
lithium metaborate, boric acid hbo2 , lithium salt, lithium metaborate, anhydrous, lithium oxido oxo borane, bo2.li, boric acid hbo2 , lithium salt 1:1, lithium metaborate,anhydrous, lithium metaborate, puratronic, lithotab boron oxide hydroxide, lithium metaborate, acs
lithium;oxido(oxo)borane
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA

SDS Documents