missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Lithium metaborate, anhydrous, 99.9% (metals basis), Alfa Aesar™

Manufacturer:  Alfa Aesar 1259130

 View more versions of this product

Catalog No. AA1259130



Specifications

Specifications

Lithium metaborate, anhydrous
99.9% (metals basis)
MFCD00011089
HZRMTWQRDMYLNW-UHFFFAOYSA-N
lithium;oxido(oxo)borane
123308
Powder
250g
13453-69-5
BLiO2
lithium metaborate, boric acid hbo2 , lithium salt, lithium metaborate, anhydrous, lithium oxido oxo borane, bo2.li, boric acid hbo2 , lithium salt 1:1, lithium metaborate,anhydrous, lithium metaborate, puratronic, lithotab boron oxide hydroxide, lithium metaborate, acs
[Li+].B(=O)[O-]
49.748
49.75
1.397g/mL

Chemical Identifiers

13453-69-5
49.748
HZRMTWQRDMYLNW-UHFFFAOYSA-N
123308
[Li+].B(=O)[O-]
BLiO2
MFCD00011089
lithium metaborate, boric acid hbo2 , lithium salt, lithium metaborate, anhydrous, lithium oxido oxo borane, bo2.li, boric acid hbo2 , lithium salt 1:1, lithium metaborate,anhydrous, lithium metaborate, puratronic, lithotab boron oxide hydroxide, lithium metaborate, acs
lithium;oxido(oxo)borane
Safety and Handling

Safety and Handling

EINECSNumber : 236-631-5

Documents