Please login to your online account to display your discounted pricing

Alfa Aesar™ Lithium metaborate, ACS, 98.0-102.0%

$115.41 - $370.66

Chemical Identifiers

Chemical Name or Material Lithium metaborate
CAS 13453-69-5
Purity Grade ACS Grade
Molecular Formula BLiO2
Linear Formula LiBO2
Formula Weight 49.75
MDL Number MFCD00011089
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3676018

  SDS
alfa aesar
3676018
50g Each for $115.41

AA3676030

  SDS
alfa aesar™
3676030
250g Each for $370.66
Description & Specifications

Specifications

Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P362-P312-P321-P405-P403+P233-P501a
Health Hazard 2 GHS H Statement
H315-H319-H335
Health Hazard 1 Warning
Physical Form Powder
Formula Weight 49.75
Purity Grade ACS Grade
Color White
Quantity 50g
Linear Formula LiBO2
Assay 98.0 to 102.0%
TSCA TSCA
Assay Percent Range 98.0-102.0%
CAS 13453-69-5
Odor Odorless
Molecular Formula BLiO2
Chemical Name or Material Lithium metaborate
MDL Number MFCD00011089
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P362-P312-P321-P405-P403+P233-P501a

GHS H Statement
H315-H319-H335