Learn More
Lithium metaborate, ACS, 98.0-102.0%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 03676030
Description
It acts as a support for antioxidant property in human and animal tissues. Used in non-linear optic materials and cellular phones. It is used for measuring total elements in soil, geological, plant and other natural organic compounds.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Lithium metaborate | |
13453-69-5 | |
Powder | |
98.0-102.0% | |
LiBO2 | |
lithium metaborate, boric acid hbo2 , lithium salt, lithium metaborate, anhydrous, lithium oxido oxo borane, bo2.li, boric acid hbo2 , lithium salt 1:1, lithium metaborate,anhydrous, lithium metaborate, puratronic, lithotab boron oxide hydroxide, lithium metaborate, acs | |
DPYKRXYVXDYLEY-UHFFFAOYSA-N | |
lithium;oxido(oxo)borane | |
123308 | |
98.0 to 102.0% | |
Odorless | |
1.397 g/mL |
845°C | |
White | |
250 g | |
B4H14Li2O7 | |
MFCD00011089 | |
soluble in ethanol. | |
[Li].[Li].[B].[B].[B].[B].O.O.O.O.O.O.O | |
183.23 | |
49.75 | |
ACS | |
Hygroscopic |
Chemical Identifiers
13453-69-5 | |
183.23 | |
DPYKRXYVXDYLEY-UHFFFAOYSA-N | |
123308 |
B4H14Li2O7 | |
MFCD00011089 | |
lithium metaborate, boric acid hbo2 , lithium salt, lithium metaborate, anhydrous, lithium oxido oxo borane, bo2.li, boric acid hbo2 , lithium salt 1:1, lithium metaborate,anhydrous, lithium metaborate, puratronic, lithotab boron oxide hydroxide, lithium metaborate, acs | |
[Li].[Li].[B].[B].[B].[B].O.O.O.O.O.O.O |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Warning
EINECSNumber : 236-631-5
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only