missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Lithium metaborate, 99%, for analysis, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 365715000
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Lithium metaborate | |
13453-69-5 | |
99% | |
LiBO2 | |
lithium metaborate, boric acid hbo2 , lithium salt, lithium metaborate, anhydrous, lithium oxido oxo borane, bo2.li, boric acid hbo2 , lithium salt 1:1, lithium metaborate,anhydrous, lithium metaborate, puratronic, lithotab boron oxide hydroxide, lithium metaborate, acs | |
HZRMTWQRDMYLNW-UHFFFAOYSA-N | |
lithium;oxido(oxo)borane | |
123308 | |
99% | |
Plastic bottle |
99% | |
500g | |
BLiO2 | |
MFCD00011089 | |
Solubility in water: 25.7g/L (20°C) | |
[Li+].B(=O)[O-] | |
49.75 | |
49.75 | |
Analysis |
Safety and Handling
EINECSNumber : 236-631-5
TSCA : TSCA
RUO â Research Use Only