missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Lithium Metaborate/Lithium Bromide (99.5%/0.50%) Fusion Flux Pure Crystal For Sample Preparation, SPEX CertiPrep™
Lithium Metaborate/Lithium Bromide (99.5%/0.50%), Pure Grade Fusion Flux for Sample Preparation, SPEX CertiPrep™. ISO Guide 34 Certified, NIST Traceable Certified Reference Materials. Supplied with comprehensive Certificate of Analysis.
Manufacturer: Spex Certiprep Llc FFB000502
Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Description
Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
- Pure Grade Fusion Flux
- 1 kg
- Lithium Metaborate/Lithium Bromide (99.5%/0.50%)
Certifications
ISO/IEC 17025:2005, ISO 17034:2016, ISO 9001: 2015
Compliance
GHS/OSHA Complaint
Specifications
Lithium Metaborate/Lithium Bromide | |
Fusion Flux Pure | |
Triple-leached LDPE bottle | |
AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation |
Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis. | |
Pure | |
1kg |
Safety and Handling
DOTInformation : Nonhazardous
Recommended Storage : Ambient
Product intended for laboratory use only