missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Lithium Met/ LiBromide 99.5%/0.50%, SPEX CertiPrep™

$875.29 - $875.29

Chemical Identifiers

Chemical Name or Material Fusion Flux Pure
Grade Pure
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
FFB000502 Spex Certiprep Llc
SPXFFB000502
Triple-leached LDPE bottle 1kg Each for $875.29
Specifications

Description

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Description & Specifications

Chemical Identifiers

Fusion Flux Pure
Pure

Specifications

1kg
Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
Lithium Metaborate/Lithium Bromide, Fusion Flux Pure
AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
Triple-leached LDPE bottle
Pure

Safety and Handling

DOTInformation : Nonhazardous

Recommended Storage : Ambient