missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Lithium L-lactate, 97%, Thermo Scientific Chemicals
$89.37 - $990.18
Chemical Identifiers
CAS | 27848-80-2 |
---|---|
Molecular Formula | C3H5LiO3 |
Molecular Weight (g/mol) | 96.01 |
MDL Number | MFCD00065399 |
InChI Key | GKQWYZBANWAFMQ-DKWTVANSSA-M |
Synonym | lithium s-2-hydroxypropanoate, l-lactic acid lithium salt, lithium l-lactate, s-2-hydroxypropionic acid lithium salt, sarcolactic acid lithium salt, lithium 1+ l-+-lactate, lithiuml-lactate, lithotab l-lactate, lactic acid lithium salt, c3h5o3.li |
PubChem CID | 23687877 |
IUPAC Name | lithium;(2S)-2-hydroxypropanoate |
SMILES | [Li+].CC(C(=O)[O-])O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1181818
|
Thermo Scientific Chemicals
A1181818 |
50 g |
N/A
|
|
|||||
AAA1181830
|
Thermo Scientific Chemicals
A1181830 |
250 g |
N/A
|
|
|||||
AAA118180B
|
Thermo Scientific Chemicals
A118180B |
1000 g |
N/A
|
|
|||||
Description
Useful chiral synthon, building block for depsipeptides
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
27848-80-2 | |
96.01 | |
GKQWYZBANWAFMQ-DKWTVANSSA-M | |
23687877 | |
[Li+].CC(C(=O)[O-])O |
C3H5LiO3 | |
MFCD00065399 | |
lithium s-2-hydroxypropanoate, l-lactic acid lithium salt, lithium l-lactate, s-2-hydroxypropionic acid lithium salt, sarcolactic acid lithium salt, lithium 1+ l-+-lactate, lithiuml-lactate, lithotab l-lactate, lactic acid lithium salt, c3h5o3.li | |
lithium;(2S)-2-hydroxypropanoate |
Specifications
27848-80-2 | |
C3H5LiO3 | |
50 g | |
Hygroscopic | |
GKQWYZBANWAFMQ-DKWTVANSSA-M | |
[Li+].CC(C(=O)[O-])O | |
96.01 | |
96.01 | |
Lithium L-lactate |
253°C to 255°C | |
MFCD00065399 | |
3568332 | |
lithium s-2-hydroxypropanoate, l-lactic acid lithium salt, lithium l-lactate, s-2-hydroxypropionic acid lithium salt, sarcolactic acid lithium salt, lithium 1+ l-+-lactate, lithiuml-lactate, lithotab l-lactate, lactic acid lithium salt, c3h5o3.li | |
−14° (c=1 in water) | |
lithium;(2S)-2-hydroxypropanoate | |
23687877 | |
97% |
Safety and Handling
EINECSNumber : 248-692-5
RTECSNumber : OJ6311000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only