Please login to your online account to display your discounted pricing

Lithium isopropoxide, pure, ACROS Organics™


Quantity 100g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC301241000

 View more versions of this product

Catalog No. AC301241000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥94% (corrected for LiOH+Li2CO3)
Formula Weight 66.03
Physical Form Powder
Quantity 100g
Color Pale yellow to beige or tan
Purity Grade Pure
Density 0.69
Packaging Glass bottle
Linear Formula LiOCH(CH3)2
Infrared Spectrum Authentic (nujol)
Health Hazard 2 GHS H Statement:
Self-heating: may catch fire.
Causes severe skin burns and eye damage.
Reacts violently with water.
MDL Number MFCD00048286
CAS 2388-10-5
Health Hazard 1 Danger
Chemical Name or Material Lithium isopropoxide
Molecular Formula C3H7LiO
Health Hazard 3 GHS P Statement:
Keep cool. Protect from sunlight.
Wear eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material Lithium isopropoxide
CAS 2388-10-5
Purity Grade Pure
Assay ≥94% (corrected for LiOH+Li2CO3)
Molecular Formula C3H7LiO
Linear Formula LiOCH(CH3)2
Formula Weight 66.03
MDL Number MFCD00048286
Accessories

Danger

GHS P Statement:
Keep cool. Protect from sunlight.
Wear eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Self-heating: may catch fire.
Causes severe skin burns and eye damage.
Reacts violently with water.

SDS & Safety Documents