missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Lithium isopropoxide, 99.9% (metals basis), Thermo Scientific Chemicals

$126.92 - $518.25

Chemical Identifiers

CAS 2388-10-5
Molecular Formula C3H7LiO
Molecular Weight (g/mol) 66.03
MDL Number MFCD00048286
InChI Key HAUKUGBTJXWQMF-UHFFFAOYSA-N
Synonym lithium isopropoxide, lithium propan-2-olate, lithium isopropanolate, lithium 2-propanolate, acmc-1ch0u, lithium isopropoxide solution, 2-propanol, lithiumsalt 1:1, lithium 1+ propan-2-olate
PubChem CID 23673350
IUPAC Name lithium;propan-2-olate
SMILES [Li+].CC(C)[O-]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA4293004
View Documents Promotion Details
Thermo Scientific Chemicals
04293004
2 g
Each for $126.92
 
AA4293009
View Documents Promotion Details
Thermo Scientific Chemicals
04293009
10 g
Each for $518.25
 
Description

Description

It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under Argon
Specifications

Chemical Identifiers

2388-10-5
66.03
HAUKUGBTJXWQMF-UHFFFAOYSA-N
23673350
[Li+].CC(C)[O-]
C3H7LiO
MFCD00048286
lithium isopropoxide, lithium propan-2-olate, lithium isopropanolate, lithium 2-propanolate, acmc-1ch0u, lithium isopropoxide solution, 2-propanol, lithiumsalt 1:1, lithium 1+ propan-2-olate
lithium;propan-2-olate

Specifications

2388-10-5
C3H7LiO
MFCD00048286
UN3206
Air and moisture sensitive
Reacts with water.
[Li+].CC(C)[O-]
66.03
66.03
Powder
(metals basis)
White to Yellow
(CH3)2CHOLi
2 g
3618614
lithium isopropoxide, lithium propan-2-olate, lithium isopropanolate, lithium 2-propanolate, acmc-1ch0u, lithium isopropoxide solution, 2-propanol, lithiumsalt 1:1, lithium 1+ propan-2-olate
HAUKUGBTJXWQMF-UHFFFAOYSA-N
lithium;propan-2-olate
23673350
99.9%
Lithium isopropoxide
Safety and Handling

Safety and Handling

GHS H Statement
H251-H314
Self-heating: may catch fire.
Causes severe skin burns and eye damage.

P210-P240-P241-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c

H228-H314-H500

DOTInformation : Hazard Class: 4.1; Packaging Group: II

TSCA : No

storageNote1 :

  • Packaged under Argon

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only