Please login to your online account to display your discounted pricing

Alfa Aesar™ Lithium isopropoxide, 94%

Chemical Identifiers

Chemical Name or Material Lithium isopropoxide
CAS 2388-10-5
Linear Formula (CH3)2CHOLi
Formula Weight 66.03
MDL Number MFCD00048286
UN Number UN3206
Beilstein 3618614
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1635203

  SDS
alfa aesar™
1635203
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1635206

  SDS
alfa aesar™
1635206
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1635214

  SDS
alfa aesar™
1635214
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 66.03
Health Hazard 3 GHS P Statement
P210-P280-P303+P361+P353-P305+P351+P338-P310-P402+P404
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.
Physical Form Powder
Linear Formula (CH3)2CHOLi
Quantity 1g
Health Hazard 1 Danger
MDL Number MFCD00048286
CAS 2388-10-5
Beilstein 3618614
Chemical Name or Material Lithium isopropoxide
UN Number UN3206
Color White
Sensitivity Air & Moisture Sensitive
Assay 94%
DOT Information Transport Hazard Class: 4.2; Packing Group: II; Proper Shipping Name: ALKALI METAL ALCOHOLATES, SELF-HEATING, CORROSIVE, N.O.S.
Health Hazard 2 GHS H Statement
H251-H314
Self-heating: may catch fire.
Causes severe skin burns and eye damage.
Safety and Handling

Danger

GHS P Statement
P210-P280-P303+P361+P353-P305+P351+P338-P310-P402+P404
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.

GHS H Statement
H251-H314
Self-heating: may catch fire.
Causes severe skin burns and eye damage.