Please login to your online account to display your discounted pricing

Lithium isopropoxide, 3M solution in THF, AcroSeal™, ACROS Organics™


Quantity 100mL
Packaging AcroSeal™ Glass Bottle

Manufacturer: thermo fisher scientific  AC396501000

 View more versions of this product

Catalog No. AC396501000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 66.03
Quantity 100mL
Packaging AcroSeal™ Glass Bottle
Concentration 3M, exact strength on the certificate of analysis
Density 0.8700g/mL
Chemical Name or Material Lithium isopropoxide
Molecular Formula CH3H7LiO
Linear Formula LiOCH(CH3)2
Appearance of Solution Clear red solution
MDL Number MFCD00048286
CAS 2388-10-5
Health Hazard 1 Danger
CAS 109-99-9
Health Hazard 3 GHS P Statement IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact
Health Hazard 2 GHS P Statement Causes severe skin burns and eye damage. May cause respiratory irritation. Highly flammable liquid and vapor. Reacts violently with water. May form explosive peroxides.

Specifications

Chemical Name or Material Lithium isopropoxide
CAS 2388-10-5
CAS 109-99-9
Molecular Formula CH3H7LiO
Linear Formula LiOCH(CH3)2
Formula Weight 66.03
MDL Number MFCD00048286
Accessories

Danger

GHS P Statement IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact

GHS P Statement Causes severe skin burns and eye damage. May cause respiratory irritation. Highly flammable liquid and vapor. Reacts violently with water. May form explosive peroxides.

SDS & Safety Documents