missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Lithium iodide, ultra dry, 99% (metals basis), Alfa Aesar™

Light Sensitive and Hygroscopic

Manufacturer:  Alfa Aesar 4415909

 View more versions of this product

Catalog No. AA4415909



Specifications

Specifications

Lithium iodide, ultra dry
99% (Metal basis)
MFCD00011092
lithium iodide, lithium iodide, lithium iodide, anhydrous, unii-s6k2xet783, lithium iodide 6lii, lithium 1+ ion iodide, lithium iodide, ultra dry, lithium iodide 7lii, lithiumiodide, lithium monoiodide
[Li+].[I-]
133.844
133.84
4.076g/cm3 at 20°C
White
10377-51-2
ILi
White powder
HSZCZNFXUDYRKD-UHFFFAOYSA-M
lithium;iodide
66321
Beads
Odorless
10g

Chemical Identifiers

10377-51-2
133.844
HSZCZNFXUDYRKD-UHFFFAOYSA-M
66321
[Li+].[I-]
ILi
MFCD00011092
lithium iodide, lithium iodide, lithium iodide, anhydrous, unii-s6k2xet783, lithium iodide 6lii, lithium 1+ ion iodide, lithium iodide, ultra dry, lithium iodide 7lii, lithiumiodide, lithium monoiodide
lithium;iodide
Safety and Handling

Safety and Handling

H315-H319-H335: Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.

EINECSNumber : 233-822-5

SDS Documents