Please login to your online account to display your discounted pricing

Alfa Aesar™ Lithium iodide, ultra dry, 99.999% (metals basis)

Chemical Identifiers

Chemical Name or Material Lithium iodide
CAS 10377-51-2
Name Note Ultra dry
Formula Weight 133.84
MDL Number MFCD00011092
Merck Index 14,5535
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1360006

 
alfa aesar™
1360006
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1360014

  SDS
alfa aesar™
1360014
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1360022

  SDS
alfa aesar™
1360022
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 133.84
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see ... on this label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
Packaging Ampule
TSCA TSCA
Physical Form Beads
Mesh Size −10 Mesh
Sensitivity Light Sensitive & Hygroscopic
Assay Percent Notes (metals basis)
CAS 10377-51-2
Storage Note 1
  • Ampouled under argon
Quantity 5g
Assay 99.999%
Health Hazard 2 GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Name Note Ultra dry
MDL Number MFCD00011092
Health Hazard 1 Warning
Refractive Index 1.955
Odor Odorless
Merck Index 14,5535
Density 4.076g/mL
Boiling Point 1170° to 1190°C
Chemical Name or Material Lithium iodide
Color White to off-white
Melting Point 449°C

  • Ampouled under argon

Safety and Handling

Warning

GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see ... on this label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.

  • Ampouled under argon