missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Lithium iodide, ultra dry, 99.999% (metals basis)

Manufacturer:  Alfa Aesar™ 1360022

 View more versions of this product

Catalog No. AA1360022



Description

Description

  • Ampouled under argon
Specifications

Specifications

Lithium iodide
10377-51-2
ILi
14,5535
HSZCZNFXUDYRKD-UHFFFAOYSA-M
lithium;iodide
66321
Beads
4.076g/mL
Odorless
1.955
1170°C to 1190°C
449°C
Ultra dry
(metals basis)
MFCD00011092
lithium iodide, lithium iodide, lithium iodide, anhydrous, unii-s6k2xet783, lithium iodide 6lii, lithium 1+ ion iodide, lithium iodide, ultra dry, lithium iodide 7lii, lithiumiodide, lithium monoiodide
[Li+].[I-]
133.844
133.84
99.999%
−10 Mesh
Ampule
Light Sensitive & Hygroscopic
White
100g

Chemical Identifiers

10377-51-2
133.844
HSZCZNFXUDYRKD-UHFFFAOYSA-M
66321
[Li+].[I-]
ILi
MFCD00011092
lithium iodide, lithium iodide, lithium iodide, anhydrous, unii-s6k2xet783, lithium iodide 6lii, lithium 1+ ion iodide, lithium iodide, ultra dry, lithium iodide 7lii, lithiumiodide, lithium monoiodide
lithium;iodide
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see ... on this label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.

Warning

TSCA : TSCA

storageNote1 :

  • Ampouled under argon

Documents