Learn More
Lithium iodide hydrate, 99%, extra pure, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 208015000
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Lithium iodide hydrate | |
500g | |
ILi·xH2O | |
MFCD00149769 | |
Solubility in water: soluble. Other solubilities: soluble in alcohol,freely soluble in acetone | |
[Li+].O.[I-] | |
133.85 | |
133.85 | |
Extra Pure |
85017-80-7 | |
98.5% min. (on dry substance) (Argentometry) | |
LiI·xH2O | |
lithium iodide hydrate, lithium iodide monohydrate, ilio, lii.h2o, lithium iodide mono hydrate, lithium iodide hydrate, puratronic, lithium 1+ ion hydrate iodide, lithium iodide lii ,monohydrate 9ci | |
WAZWGFFJLSIDMX-UHFFFAOYSA-M | |
lithium;iodide;hydrate | |
23688290 | |
99% | |
Glass bottle |
Safety and Handling
GHS H Statement
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at
GHS Signal Word: Warning
RUO â Research Use Only