Learn More
Lithium iodide hydrate, 99%, extra pure, Thermo Scientific Chemicals
$202.97 - $202.97
Chemical Identifiers
CAS | 85017-80-7 |
---|---|
Molecular Formula | ILi |
Molecular Weight (g/mol) | 133.84 |
MDL Number | MFCD00149769 |
InChI Key | HSZCZNFXUDYRKD-UHFFFAOYSA-M |
Synonym | lithium iodide hydrate, lithium iodide monohydrate, ilio, lii.h2o, lithium iodide mono hydrate, lithium iodide hydrate, puratronic, lithium 1+ ion hydrate iodide, lithium iodide lii ,monohydrate 9ci |
PubChem CID | 23688290 |
SMILES | [Li+].[I-] |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC208011000
|
Thermo Scientific Chemicals
208011000 |
100 g | Glass bottle |
Each for $202.97
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
85017-80-7 | |
133.84 | |
HSZCZNFXUDYRKD-UHFFFAOYSA-M | |
23688290 |
ILi | |
MFCD00149769 | |
lithium iodide hydrate, lithium iodide monohydrate, ilio, lii.h2o, lithium iodide mono hydrate, lithium iodide hydrate, puratronic, lithium 1+ ion hydrate iodide, lithium iodide lii ,monohydrate 9ci | |
[Li+].[I-] |
Specifications
1171°C | |
85017-80-7 | |
Crystalline Powder and/or Crystals | |
98.5% min. (on dry substance) (Argentometry) | |
LiI·xH2O | |
lithium iodide hydrate, lithium iodide monohydrate, ilio, lii.h2o, lithium iodide mono hydrate, lithium iodide hydrate, puratronic, lithium 1+ ion hydrate iodide, lithium iodide lii ,monohydrate 9ci | |
HSZCZNFXUDYRKD-UHFFFAOYSA-M | |
lithium;iodide;hydrate | |
23688290 | |
99% | |
Glass bottle |
73°C | |
White to Yellow | |
100 g | |
ILi | |
MFCD00149769 | |
Solubility in water: soluble. Other solubilities: soluble in alcohol,freely soluble in acetone | |
[Li+].[I-] | |
133.84 | |
133.85 | |
Extra Pure | |
Lithium iodide hydrate |
Safety and Handling
GHS H Statement
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at
GHS Signal Word: Warning
RUO – Research Use Only