missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Lithium iodide hydrate, 99%, extra pure, ACROS Organics™

Manufacturer:  Acros Organics 208011000

 View more versions of this product

Catalog No. AC208011000



Specifications

Specifications

Lithium iodide hydrate
99%
LiI·xH2O
ilio
WAZWGFFJLSIDMX-UHFFFAOYSA-M
lithium;iodide;hydrate
23688290
Crystals or Crystalline Powder
Extra Pure
10ppm max.
1000ppm max.
1171°C
73°C
85017-80-7
ILi·xH2O
MFCD00149769
Solubility in water: soluble. Other solubilities: soluble in alcohol,freely soluble in acetone
[Li+].O.[I-]
133.85
133.85
≥98.5% (on dry substance)
15ppm max.
Glass bottle
30% max. (K.F.)
White to Yellow
100g

Chemical Identifiers

85017-80-7
133.85
WAZWGFFJLSIDMX-UHFFFAOYSA-M
23688290
[Li+].O.[I-]
ILi·xH2O
MFCD00149769
ilio
lithium;iodide;hydrate
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS Documents