Please login to your online account to display your discounted pricing

Alfa Aesar™ Lithium ingot, 5.7cm (2.2 in.) dia. x 8.6cm (3.4 in.) long, 99.9% (metals basis)

Chemical Identifiers

Chemical Name or Material Lithium ingot
CAS 7439-93-2
Name Note 5.7cm (2.2 in.) dia. x 8.6cm (3.4 in.) long
MDL Number MFCD00134051
UN Number UN1415
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA00658KF

  SDS
alfa aesar™
00658KF
1 piece CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA00658KI

  SDS
alfa aesar™
00658KI
4 pieces CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA00658P0

  SDS
alfa aesar™
00658P0
4 x 1 piece CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Lithium ingot
TSCA TSCA
Name Note 5.7cm (2.2 in.) dia. x 8.6cm (3.4 in.) long
Physical Form Pieces
Assay 99.9%
Assay Percent Notes (metals basis)
MDL Number MFCD00134051
CAS 7439-93-2
UN Number UN1415
Odor Odorless
Packaging Vacuum can
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H260-H314
In contact with water releases flammable gases which may ignite spontaneously.
Causes severe skin burns and eye damage.
Health Hazard 3 GHS P Statement
P231+P232-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Handle under inert gas.
Protect from moisture.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Storage Note 1
  • In vacuum can with mineral oil coating
Color Silver-white
DOT Information Transport Hazard Class: 4.3; Packing Group: I; Proper Shipping Name: LITHIUM
RTECS Number OJ5540000
Weight ≈116g/pc
Quantity 1 piece

  • In vacuum can with mineral oil coating

Safety and Handling

Danger

GHS P Statement
P231+P232-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Handle under inert gas.
Protect from moisture.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H260-H314
In contact with water releases flammable gases which may ignite spontaneously.
Causes severe skin burns and eye damage.

  • In vacuum can with mineral oil coating