missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing
Lithium ICP Standard, 1000 ppm Li in 3% HNO3, Ricca Chemical
Chemical Identifiers
CAS |
554-13-2 |
Molecular Formula |
CLi2O3 |
Molecular Weight (g/mol) |
73.89 |
MDL Number |
MFCD00011084 |
InChI Key |
XGZVUEUWXADBQD-UHFFFAOYSA-L |
PubChem CID |
11125 |
ChEBI |
CHEBI:6504 |
IUPAC Name |
dilithium(1+) carbonate |
SMILES |
[Li+].[Li+].[O-]C([O-])=O |
View More Specs
Products
1
Description
Specifications
Chemical Identifiers
|
554-13-2 |
|
73.89 |
|
XGZVUEUWXADBQD-UHFFFAOYSA-L |
|
CHEBI:6504 |
|
[Li+].[Li+].[O-]C([O-])=O |
|
CLi2O3 |
|
MFCD00011084 |
|
11125 |
|
dilithium(1+) carbonate |
Specifications
|
554-13-2,7697-37-2,554-13-2,7732-18-5 |
|
100 mL |
|
96.0,5.62,0.51 |
|
1,000ppm Li |
|
MFCD00011084 |
|
[Li+].[Li+].[O-]C([O-])=O |
|
73.89 |
|
CHEBI:6504 |
|
Liquid |
|
Natural LDPE Bottle |
|
92.0,5.4,0.49 |
|
Colorless |
|
CLi2O3 |
|
XGZVUEUWXADBQD-UHFFFAOYSA-L |
|
dilithium(1+) carbonate |
|
11125 |
|
1 mL = 1 mg Li |
|
Lithium ICP Standard |
SDS
missing translation for 'documents'