Learn More
Lithium hydroxide monohydrate, 99.995% (metals basis), Thermo Scientific Chemicals
Lithium hydroxide monohydrate, H3LiO2, CAS Number-1310-66-3, unii-g51xlp968g, lithium hydroxide monohydrate, lioh water, lithiumhydrate, hydroxyde de lithium, lithium hydroxido, lithium hydroxide, monohydrate, lithium hydroxide hydrate, lioh.hydrate, lioh-hydrate, 100g, 99.995% (Metals basis), White
$178.96 - $2386.21
Chemical Identifiers
CAS | 1310-66-3 |
---|---|
Molecular Formula | H3LiO2 |
Molecular Weight (g/mol) | 41.96 |
MDL Number | MFCD00149772 |
InChI Key | GLXDVVHUTZTUQK-UHFFFAOYSA-M |
Synonym | lithium hydroxide monohydrate, lithium hydroxide hydrate, lithium hydroxido, hydroxyde de lithium, unii-g51xlp968g, lithium hydroxide, monohydrate, lithiumhydrate, lioh-hydrate, lioh.hydrate, lioh water |
PubChem CID | 168937 |
IUPAC Name | lithium;hydroxide;hydrate |
SMILES | [Li+].O.[OH-] |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AA4447314
|
Thermo Scientific Chemicals
04447314 |
25 g |
Each for $178.96
|
|
|||||
AA4447322
|
Thermo Scientific Chemicals
04447322 |
100 g |
Each for $533.44
|
|
|||||
AA4447336
|
Thermo Scientific Chemicals
04447336 |
500 g |
Each for $2,386.21
|
|
|||||
Description
Lithium hydroxide is used for the production of lithium greases, lithium soaps, lithium stearate and lithium salts. It finds application as a carbon dioxide adsorbent in breathing gas purification systems for spacecrafts, submarines and rebreathers; as a storage-battery electrolyte; as a heat transfer medium and as a catalyst for polymerization reaction. It is also used in ceramics and some portland cement formulations.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1310-66-3 | |
41.96 | |
GLXDVVHUTZTUQK-UHFFFAOYSA-M | |
168937 | |
[Li+].O.[OH-] |
H3LiO2 | |
MFCD00149772 | |
lithium hydroxide monohydrate, lithium hydroxide hydrate, lithium hydroxido, hydroxyde de lithium, unii-g51xlp968g, lithium hydroxide, monohydrate, lithiumhydrate, lioh-hydrate, lioh.hydrate, lioh water | |
lithium;hydroxide;hydrate |
Specifications
1310-66-3 | |
Powder | |
99.995% (Metals basis) | |
MFCD00149772 | |
Soluble in water,methanol and ethanol. | |
[Li+].O.[OH-] | |
41.96 | |
41.96 (23.95 Anhydrous) | |
Lithium hydroxide monohydrate |
White | |
25 g | |
H3LiO2 | |
lithium hydroxide monohydrate, lithium hydroxide hydrate, lithium hydroxido, hydroxyde de lithium, unii-g51xlp968g, lithium hydroxide, monohydrate, lithiumhydrate, lioh-hydrate, lioh.hydrate, lioh water | |
GLXDVVHUTZTUQK-UHFFFAOYSA-M | |
lithium;hydroxide;hydrate | |
168937 | |
1.51 g/cm3 at 20°C |
Safety and Handling
Keep container in a well ventilated place. When using do not eat or drink. In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable protective clothing, gloves and eye/face protection. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible). This material and/or its container must be disposed of as hazardous waste.
P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
H302-H314-H335
DOTInformation : DOT Class: 8, Packaging Group: II
EINECSNumber : 215-183-4
RTECSNumber : OJ6307080
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only