Learn More
Lithium hydroxide monohydrate, extra pure, ca. 56% LiOH, Thermo Scientific Chemicals
$48.01 - $782.05
Chemical Identifiers
CAS | 1310-66-3 |
---|---|
Molecular Formula | H3LiO2 |
Molecular Weight (g/mol) | 41.96 |
MDL Number | MFCD00149772 |
InChI Key | GLXDVVHUTZTUQK-UHFFFAOYSA-M |
Synonym | lithium hydroxide monohydrate, lithium hydroxide hydrate, lithium hydroxido, hydroxyde de lithium, unii-g51xlp968g, lithium hydroxide, monohydrate, lithiumhydrate, lioh-hydrate, lioh.hydrate, lioh water |
PubChem CID | 168937 |
SMILES | [Li+].O.[OH-] |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC199540250
|
Thermo Scientific Chemicals
199540250 |
25 g | Plastic bottle |
Each for $48.01
|
|
||||
AC199542500
|
Thermo Scientific Chemicals
199542500 |
250 g | Plastic bottle |
Each for $216.60
|
|
||||
AC199540010
|
Thermo Scientific Chemicals
199540010 |
1 kg | Plastic bottle |
Each for $782.05
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1310-66-3 | |
41.96 | |
GLXDVVHUTZTUQK-UHFFFAOYSA-M | |
168937 |
H3LiO2 | |
MFCD00149772 | |
lithium hydroxide monohydrate, lithium hydroxide hydrate, lithium hydroxido, hydroxyde de lithium, unii-g51xlp968g, lithium hydroxide, monohydrate, lithiumhydrate, lioh-hydrate, lioh.hydrate, lioh water | |
[Li+].O.[OH-] |
Specifications
920.0°C | |
1310-66-3 | |
Crystalline Powder | |
H3LiO2 | |
MFCD00149772 | |
14,5534 | |
Solubility in water: 109g/L (20°C). Other solubilities: 107g/L (0°C), 148g/L (100°C), slightly soluble in alcohol | |
[Li+].O.[OH-] | |
41.96 | |
41.95 | |
Extra Pure | |
Lithium hydroxide monohydrate |
462.0°C | |
Cream to White or Yellow | |
25 g | |
LiOH·H2O | |
07,208; 12,282 | |
lithium hydroxide monohydrate, lithium hydroxide hydrate, lithium hydroxido, hydroxyde de lithium, unii-g51xlp968g, lithium hydroxide, monohydrate, lithiumhydrate, lioh-hydrate, lioh.hydrate, lioh water | |
GLXDVVHUTZTUQK-UHFFFAOYSA-M | |
lithium(1+) hydrate hydroxide | |
168937 | |
∼56% (LiOH) | |
Plastic bottle |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Causes severe skin burns and eye damage.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre
GHS Signal Word: Danger
RUO – Research Use Only