Please login to your online account to display your discounted pricing

Lithium Hydroxide, Monohydrate, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

High quality chemicals for laboratory and specialized industrial use

$309.14 - $413.36

Chemical Identifiers

Chemical Name or Material Lithium Hydroxide
CAS 1310-66-3
Name Note Monohydrate
Purity Grade Baker Analyzed™ ACS Reagent
Molecular Formula LiOH·H2O
Formula Weight 41.96
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02003216

 
avantor performance
P40608
Poly Bottle 1kg Each for $413.36

02003217

 
avantor performance
P40607
Poly Bottle 500g Each for $309.14
Description & Specifications

Specifications

Purity Grade Baker Analyzed™ ACS Reagent
Formula Weight 41.96
Molecular Formula LiOH·H2O
CAS 1310-66-3
Chemical Name or Material Lithium Hydroxide
Name Note Monohydrate
Packaging Poly Bottle
Specific Gravity 1L = 1.51kg
Quantity 1kg