Please login to your online account to display your discounted pricing

Lithium Hydroxide, Monohydrate, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™


High quality chemicals for laboratory and specialized industrial use

Quantity 500g
Packaging Poly Bottle

Manufacturer: avantor performance  P40607

 View more versions of this product

Catalog No. 02003217

  • / Each
 

Description & Specifications

Chemical Identifiers

Purity Grade Baker Analyzed™ ACS Reagent
Formula Weight 41.96
Chemical Name or Material Lithium Hydroxide
Molecular Formula LiOH·H2O
CAS 1310-66-3
Name Note Monohydrate
Packaging Poly Bottle
Specific Gravity 1L = 1.51kg
Quantity 500g

Specifications

Chemical Name or Material Lithium Hydroxide
CAS 1310-66-3
Name Note Monohydrate
Purity Grade Baker Analyzed™ ACS Reagent
Molecular Formula LiOH·H2O
Formula Weight 41.96