Please login to your online account to display your discounted pricing

Alfa Aesar™ Lithium hydroxide monohydrate, 98%

$29.38 - $29.38

Chemical Identifiers

Chemical Name or Material Lithium hydroxide monohydrate
CAS 1310-66-3
Molecular Formula LiOH·H2O
Formula Weight 41.96 (23.95 Anhydrous)
MDL Number MFCD00149772
UN Number UN2680
Merck Index 14,5534
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA155190E

  SDS
alfa aesar™
A155190E
2500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1551922

  SDS
alfa aesar™
A1551922
100g Each for $29.38

AAA1551936

  SDS
alfa aesar™
A1551936
500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 41.96 (23.95 Anhydrous)
Health Hazard 3 GHS P Statement
P260-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
RTECS Number OJ6307080
DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: LITHIUM HYDROXIDE
Chemical Name or Material Lithium hydroxide monohydrate
Health Hazard 1 Danger
Assay 98%
CAS 1310-66-3
UN Number UN2680
Health Hazard 2 GHS H Statement
H301-H331-H314-H318
Toxic if swallowed.
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Molecular Formula LiOH·H2O
MDL Number MFCD00149772
TSCA TSCA
Merck Index 14,5534
Quantity 2500g

Applications
Lithium hydroxide is used for the production of lithium greases, lithium soaps, lithium stearate and lithium salts. It finds application as a carbon dioxide adsorbent in breathing gas purification systems for spacecrafts, submarines and rebreathers; as a storage-battery electrolyte; as a heat transfer medium and as a catalyst for polymerization reaction. It is also used in ceramics and some portland cement formulations.

Solubility
Soluble in water, methanol and ethanol.

Notes
Air sensitive and hygroscopic. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with moisture, strong oxidizing agents, strong acids and carbon dioxide.

Safety and Handling

Danger

GHS P Statement
P260-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H331-H314-H318
Toxic if swallowed.
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.