missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Lithium hydroxide, anhydrous, 99.995% (metals basis)

Manufacturer:  Alfa Aesar™ 4447106

 View more versions of this product

Catalog No. AA4447106



Specifications

Specifications

Lithium hydroxide
1310-65-2
HLiO
UN2680
lioh
WMFOQBRAJBCJND-UHFFFAOYSA-M
lithium;hydroxide
3939
23.95
99.995%
Odorless
924°C
462°C
5g
Anhydrous
(metals basis)
MFCD00011095
14,5534
Soluble in water. Slightly soluble in ethanol and methanol
[Li+].[OH-]
23.947
CHEBI:33979
Powder
2.54 g/mL
Air sensitive; Hygroscopic
White
14

Chemical Identifiers

1310-65-2
23.947
WMFOQBRAJBCJND-UHFFFAOYSA-M
3939
lithium;hydroxide
HLiO
MFCD00011095
lioh
CHEBI:33979
[Li+].[OH-]
Safety and Handling

Safety and Handling

GHS H Statement
H301-H331-H314-H318
Toxic if swallowed.
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c

H302-H314-H335-H336-H373

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: LITHIUM HYDROXIDE

EINECSNumber : 215-183-4

RTECSNumber : OJ6307070

TSCA : Yes

Recommended Storage : Ambient temperatures; Store under Nitrogen

Documents