missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Lithium hydroxide, anhydrous, 98%, Alfa Aesar™

Manufacturer:  Alfa Aesar 1340736

 View more versions of this product

Catalog No. AA1340736



Specifications

Specifications

Lithium hydroxide, anhydrous
98%
MFCD00011095
lithium hydroxide, lithium hydrate, lioh, lithium hydroxide anhydrous, lithium hydroxide li oh, lithiumhydroxid, lithiumhydroxide, lithium hydoxide, unii-903yl31jas, lithium hydroxide, anhydrous
[Li+].[OH-]
23.947
CHEBI:33979
4+14 Mesh
Odorless
500g
1310-65-2
HLiO
White powder
WMFOQBRAJBCJND-UHFFFAOYSA-M
lithium;hydroxide
3939
23.95
2.54g/cm3 at 20°C
White

Chemical Identifiers

1310-65-2
23.947
WMFOQBRAJBCJND-UHFFFAOYSA-M
3939
lithium;hydroxide
HLiO
MFCD00011095
lithium hydroxide, lithium hydrate, lioh, lithium hydroxide anhydrous, lithium hydroxide li oh, lithiumhydroxid, lithiumhydroxide, lithium hydoxide, unii-903yl31jas, lithium hydroxide, anhydrous
CHEBI:33979
[Li+].[OH-]
Safety and Handling

Safety and Handling

H301-H331-H314-H318: Toxic if swallowed. Toxic if inhaled. Causes severe skin burns and eye damage. Causes serious eye damage.

DOTInformation : DOT Class: 8, Packaging Group: II

EINECSNumber : 215-183-4

Documents