Please login to your online account to display your discounted pricing

Alfa Aesar™ Lithium hydride, 97+%

Chemical Identifiers

Chemical Name or Material Lithium hydride
CAS 7580-67-8
Molecular Formula HLi
Linear Formula LiH
Formula Weight 7.95
MDL Number MFCD00011074
UN Number UN1414
Merck Index 14,5533
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1794822

  SDS
alfa aesar™
A1794822
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1794818

  SDS
alfa aesar™
A1794818
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1794830

  SDS
alfa aesar™
A1794830
250g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 7.95
Chemical Name or Material Lithium hydride
TSCA TSCA
Odor Odorless
Assay ≥97%
Molecular Formula HLi
Linear Formula LiH
MDL Number MFCD00011074
CAS 7580-67-8
UN Number UN1414
Merck Index 14,5533
Melting Point 688°C
Quantity 100g
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H260-H301-H314-H318
In contact with water releases flammable gases which may ignite spontaneously.
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3 GHS P Statement
P231+P232-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
Handle under inert gas.
Protect from moisture.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 4.3; Packing Group: I; Proper Shipping Name: LITHIUM HYDRIDE
RTECS Number OJ6300000

Applications
Used for preparation of anhydrous Li salts of carboxylic acids prior to reaction with organothium reagents (most often methyl or phenyl) as a useful route to ketones. Also used in the preparation of LiCN by reaction with acetone cyanohydrins. It is also applied as a desiccating and condensing agent and a shielding material in nuclear energy production. It is used in the manufacture of catalysts for polyolefin plastics; in fuels for aircraft, missiles. It is used in metallurgy, as degasifier, deoxidizer, desulfurizer; as grain refiner in non-ferrous metals. Employed as a chemical intermediate for butyllithium polymerization catalyst; in alloys with aluminum or magnesium for aerospace uses.

Solubility
Reacts violently with water. Soluble in ether, liquid ammonia. Insoluble in benzene, and toluene.

Notes
Air & Moisture Sensitive. Store away from acids, alcohols, oxidizing agents, air, water/moisture. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.

Safety and Handling

Danger

GHS P Statement
P231+P232-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
Handle under inert gas.
Protect from moisture.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H260-H301-H314-H318
In contact with water releases flammable gases which may ignite spontaneously.
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.