missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Lithium Hexafluorophosphate, TCI America™

Manufacturer:  TCI America L014625G

 View more versions of this product

Catalog No. L014625G



Specifications

Specifications

Lithium Hexafluorophosphate
200°C
21324-40-3
MFCD00011096
lithium hexafluorophosphate, lithium hexafluorophosphate v, phosphate 1-, hexafluoro-, lithium, phosphate 1-, hexafluoro-, lithium 1:1, lithiumhexafluorophosphate, lithium hexafluorophosphate 1-, lipf6, lithium hexafluoroposphate, acmc-209fj7, ksc492g4h
[Li+].F[P-](F)(F)(F)(F)F
151.904
151.90
≥97.0% (T)
White
25g
F6LiP
2923
AXPLOJNSKRXQPA-UHFFFAOYSA-N
lithium;hexafluorophosphate
23688915
Crystal-Powder at 20°C

Chemical Identifiers

21324-40-3
151.904
AXPLOJNSKRXQPA-UHFFFAOYSA-N
23688915
[Li+].F[P-](F)(F)(F)(F)F
F6LiP
MFCD00011096
lithium hexafluorophosphate, lithium hexafluorophosphate v, phosphate 1-, hexafluoro-, lithium, phosphate 1-, hexafluoro-, lithium 1:1, lithiumhexafluorophosphate, lithium hexafluorophosphate 1-, lipf6, lithium hexafluoroposphate, acmc-209fj7, ksc492g4h
lithium;hexafluorophosphate
Safety and Handling

Safety and Handling

TSCA : Yes

Documents