missing translation for 'onlineSavingsMsg'
Learn More

Lithium hexafluoroarsenate(V), 99%

CAS: 29935-35-1 | AsF6Li | 195.85 g/mol

$278.92 - $1124.49

Chemical Identifiers

CAS 29935-35-1
Molecular Formula AsF6Li
Molecular Weight (g/mol) 195.85
MDL Number MFCD00011076
InChI Key YYPUQABLGAAHDV-UHFFFAOYSA-H
Synonym lithium hexafluoroarsenate, lithium hexafluoroarsenate v, lithium hexafluoroarsoranuide, lithium hexakis fluoranyl arsenic 1-, lithotab hexafluoro-$l^ 5-arsanuide, lithium 1+ hexafluoro-??-arsanuide
PubChem CID 9837036
SMILES [Li+].[F-].[F-].[F-].[F-].[F-].[F-].[As+5]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA1153009
View Documents
Thermo Scientific Chemicals
01153009
10 g
Each for $278.92
Only null left
 
AA1153018
View Documents
Thermo Scientific Chemicals
01153018
50 g
Each for $1,124.49
Only null left
 
Description

Description

Lithium hexafluoroarsenate(V) is a potential battery electrolyte material.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

29935-35-1
195.85
YYPUQABLGAAHDV-UHFFFAOYSA-H
9837036
AsF6Li
MFCD00011076
lithium hexafluoroarsenate, lithium hexafluoroarsenate v, lithium hexafluoroarsoranuide, lithium hexakis fluoranyl arsenic 1-, lithotab hexafluoro-$l^ 5-arsanuide, lithium 1+ hexafluoro-??-arsanuide
[Li+].[F-].[F-].[F-].[F-].[F-].[F-].[As+5]

Specifications

29935-35-1
10 g
MFCD00011076
lithium hexafluoroarsenate, lithium hexafluoroarsenate v, lithium hexafluoroarsoranuide, lithium hexakis fluoranyl arsenic 1-, lithotab hexafluoro-$l^ 5-arsanuide, lithium 1+ hexafluoro-??-arsanuide
YYPUQABLGAAHDV-UHFFFAOYSA-H
lithium;hexafluoroarsenic(1-)
9837036
99%
Lithium hexafluoroarsenate(V)
Powder
AsF6Li
UN1557
Insoluble in water.
[Li+].[F-].[F-].[F-].[F-].[F-].[F-].[As+5]
195.85
195.85
Hygroscopic
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H331
Toxic if swallowed.
Toxic if inhaled.

P201-P202-P261-P264b-P270-P271-P281-P301+P310-P304+P340-P308+P313-P311-P330-P501c

H301+H331-H350

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: ARSENIC COMPOUNDS, SOLID, N.O.S.

EINECSNumber : 249-963-0

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only