missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Lithium Formate Monohydrate, TCI America™

Manufacturer:  TCI America F017125G

 View more versions of this product

Catalog No. F017125G



Specifications

Specifications

Lithium Formate Monohydrate
25g
CH3LiO3
lithium formate monohydrate, unii-zqs5s890b6, formic acid, lithium salt, monohydrate, lithium formate hydrate, monolithium formate monohydrate, lithium formate monohydrate mi, acmc-209mow, lithotab formira hydrate, li.cho2.h2o, lithium formate, reagent
[Li+].C(=O)[O-].O
69.972
51.96
≥98.0% (T)
White
6108-23-2
MFCD00050823
RDAKCPVJSPEDON-UHFFFAOYSA-M
lithium;formate;hydrate
23702287
Crystal-Powder at 20°C

Chemical Identifiers

6108-23-2
69.972
RDAKCPVJSPEDON-UHFFFAOYSA-M
23702287
[Li+].C(=O)[O-].O
CH3LiO3
MFCD00050823
lithium formate monohydrate, unii-zqs5s890b6, formic acid, lithium salt, monohydrate, lithium formate hydrate, monolithium formate monohydrate, lithium formate monohydrate mi, acmc-209mow, lithotab formira hydrate, li.cho2.h2o, lithium formate, reagent
lithium;formate;hydrate
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0676

TSCA : Yes

Documents