missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Lithium foil, 99.9% (metals basis), Alfa Aesar™

Manufacturer:  Thermo Fisher Scientific 1076722

Catalog No. AA1076722



Specifications

Specifications

Lithium foil
7439-93-2
Li
hsdb 549
[Li]
6.94
CHEBI:30145
100g
99.9% (metals basis)
MFCD00134051
WHXSMMKQMYFTQS-UHFFFAOYSA-N
lithium
3028194
≈1.2m/100g,Packaged under argon

Chemical Identifiers

7439-93-2
6.94
WHXSMMKQMYFTQS-UHFFFAOYSA-N
3028194
lithium
Li
MFCD00134051
hsdb 549
CHEBI:30145
[Li]
Safety and Handling

Safety and Handling

P223-P231+P232-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P335+P334-P363-P370+P378q-P501c

H260-H314

DOTInformation : Hazard Class: 4.3; Packaging Group: I

EINECSNumber : 231-102-5

RTECSNumber : OJ5540000

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents